Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003817.3 | NP_003808.2 | 2 | 241 | ||
XM_017013943.1 | XP_016869432.1 | 2 | 128 | |||
GenBank mRNA | AF215824.1 | - | 2 | 212 | ||
AK292492.1 | - | 2 | 245 | |||
BC058037.1 | - | 2 | 310 | |||
DB077360.1 | - | 2 | 245 |