Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAALAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300930.1 | NP_001287859.1 | 6 | 807 | ||
NM_005467.3 | NP_005458.1 | 6 | 807 | |||
XM_017017043.1 | XP_016872532.1 | 8 | 973 | |||
XM_017017044.1 | XP_016872533.1 | 8 | 974 | |||
XM_017017045.1 | XP_016872534.1 | 6 | 810 | |||
XM_017017046.1 | XP_016872535.1 | 8 | 974 | |||
GenBank mRNA | AJ012370.1 | - | 6 | 743 | ||
AK075390.1 | - | 6 | 782 | |||
BC027594.1 | - | 6 | 763 | |||
BC038840.1 | - | 6 | 793 | |||
BC096316.3 | - | 6 | 719 | 40007638 | ||
BC096317.3 | - | 6 | 718 | 40007639 | ||
BC096318.3 | - | 6 | 718 | 40007641 | ||
BC099646.4 | - | 6 | 719 |