Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENDOU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172439.1 | NP_001165910.1 | 4 | 467 | ||
NM_001172440.1 | NP_001165911.1 | 2 | 278 | |||
NM_006025.3 | NP_006016.1 | 3 | 344 | |||
GenBank mRNA | AK075446.1 | - | 4 | 431 | ||
AK295963.1 | - | 1 | 66 | |||
AK300169.1 | - | 2 | 240 | |||
AK302308.1 | - | 1 | 58 | |||
AK314688.1 | - | 3 | 214 | |||
AX772808.1 | - | 4 | 419 | |||
BC069715.1 | - | 3 | 223 | 7262649 | ||
BC074763.2 | - | 3 | 229 | 30915175 | ||
BC111782.1 | - | 3 | 318 | 40037008 | ||
DA838842.1 | - | 3 | 345 | |||
DA841959.1 | - | 3 | 344 | |||
DC381238.1 | - | 2 | 279 | |||
M32402.1 | - | 3 | 339 | |||
M36109.1 | - | 3 | 339 |