Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006988.4 | NP_008919.3 | 2 | 1234 | ||
GenBank mRNA | AB037767.1 | - | 2 | 894 | ||
AF060152.1 | - | 2 | 965 | |||
AF170084.1 | - | 2 | 1021 | |||
AF207664.1 | - | 2 | 1221 | |||
AK129873.1 | - | 2 | 85 | |||
AK314475.1 | - | 2 | 1234 | |||
BC036515.2 | - | 2 | 1234 | 5266513 | ||
BX647388.1 | - | 2 | 1233 |