Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012128.3 | NP_036260.2 | 2 | 227 | ||
NR_024602.1 | 2 | 227 | ||||
XM_011541015.2 | XP_011539317.1 | 2 | 46 | |||
GenBank mRNA | AF127035.1 | - | 2 | 211 | ||
AK000072.1 | - | 2 | 225 | |||
AK293766.1 | - | 2 | 60 | |||
AK307785.1 | - | 2 | 171 | |||
AK307898.1 | - | 2 | 88 | |||
AY358470.1 | - | 2 | 207 | |||
BC113687.1 | - | 2 | 222 | 8322739 | ||
BC113689.1 | - | 2 | 221 | 8322741 | ||
DA668017.1 | - | 2 | 227 | |||
DA761219.1 | - | 2 | 289 |