Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197941.3 | NP_922932.2 | 7 | 1915 | ||
NR_135689.1 | 7 | 1920 | ||||
XM_011543113.2 | XP_011541415.1 | 7 | 1930 | |||
XM_011543114.2 | XP_011541416.1 | 7 | 1509 | |||
XM_011543115.2 | XP_011541417.1 | 7 | 1968 | |||
XM_011543116.2 | XP_011541418.1 | 7 | 1965 | |||
XM_011543117.1 | XP_011541419.1 | 5 | 1268 | |||
XM_011543119.2 | XP_011541421.1 | 7 | 1959 | |||
XM_011543120.2 | XP_011541422.1 | 7 | 1956 | |||
XM_011543121.2 | XP_011541423.2 | 3 | 296 | |||
XM_011543122.2 | XP_011541424.1 | 7 | 1956 | |||
XM_011543126.2 | XP_011541428.1 | 7 | 1953 | |||
XM_011543127.2 | XP_011541429.1 | 7 | 1953 | |||
GenBank mRNA | AB209685.1 | - | 7 | 1417 | ||
AF140674.1 | - | 6 | 1079 | |||
AY692424.1 | - | 7 | 1873 | |||
AY692425.1 | - | 6 | 1077 | |||
AY692426.1 | - | 6 | 1081 |