Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167595.1 | NP_001161067.1 | 5 | 1202 | ||
NM_014324.5 | NP_055139.4 | 5 | 1202 | |||
NM_203382.2 | NP_976316.1 | 4 | 1041 | |||
GenBank mRNA | AF047020.1 | - | 5 | 1194 | ||
AF158378.1 | - | 5 | 1171 | |||
AJ130733.1 | - | 5 | 1194 | |||
AK000912.1 | - | 5 | 1187 | |||
AK222831.1 | - | 5 | 1174 | |||
AK225749.1 | - | 5 | 1211 | |||
AY935981.1 | - | 5 | 1115 | |||
BC009471.1 | - | 4 | 957 | 2958112 | ||
BQ941482.1 | - | 3 | 711 | |||
EF560721.1 | - | 5 | 1125 | |||
FJ498906.1 | - | 5 | 1000 | |||
FJ498907.1 | - | 5 | 1000 | |||
FJ498908.1 | - | 5 | 1130 |
Gene Symbol: C1QTNF3-AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037951.1 | 9 | 1462 | |||
GenBank mRNA | AB209261.1 | - | 9 | 1471 |