Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015143.2 | NP_055958.2 | 11 | 1538 | ||
XM_011531777.2 | XP_011530079.1 | 11 | 1535 | |||
XM_011531778.1 | XP_011530080.1 | 11 | 1331 | |||
XM_011531779.1 | XP_011530081.1 | 10 | 1406 | |||
XM_017007914.1 | XP_016863403.1 | 10 | 1394 | |||
XM_017007915.1 | XP_016863404.1 | 12 | 1552 | |||
XM_017007916.1 | XP_016863405.1 | 11 | 1551 | |||
XM_017007917.1 | XP_016863406.1 | 11 | 1327 | |||
GenBank mRNA | AK309465.1 | - | 3 | 611 | ||
BC030054.1 | - | 7 | 1096 | 3958447 | ||
D42084.1 | - | 11 | 1446 |