Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016246.2 | NP_057330.2 | 5, 6 | 627 | ||
XM_005258969.3 | XP_005259026.1 | 5, 6 | 636 | |||
GenBank mRNA | AF126781.1 | - | 5, 6 | 473 | ||
AK307437.1 | - | 5, 6 | 436 | |||
AY358430.1 | - | 5, 6 | 466 | |||
BC006283.2 | - | 5, 6 | 420 | 4026397 | ||
BC006294.1 | - | 5, 6 | 428 | 4025809 | ||
BI602824.1 | - | 5 | 633 |