Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016246.2 | NP_057330.2 | 8 | 863 | ||
GenBank mRNA | AF126781.1 | - | 8 | 709 | ||
AY358430.1 | - | 8 | 702 | |||
BC006283.2 | - | 8 | 656 | 4026397 | ||
BC006294.1 | - | 8 | 664 | 4025809 |