Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083907.2 | NP_001077376.2 | 5 | 875 | ||
NM_001127507.2 | NP_001120979.2 | 4 | 707 | |||
NM_017935.4 | NP_060405.4 | 5 | 1106 | |||
XM_017008337.1 | XP_016863826.1 | 5 | 853 | |||
GenBank mRNA | AB063170.1 | - | 4 | 742 | ||
AK000713.1 | - | 5 | 822 | |||
AK091523.1 | - | 9 | 1207 | |||
AK292133.1 | - | 5 | 950 | |||
AL834291.1 | - | 5 | 925 | |||
BC032241.1 | - | 5 | 974 | 5214147 | ||
EU051376.1 | - | 4 | 445 |