Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004360.3 | NP_001004360.2 | 9 | 948 | ||
NM_001178034.1 | NP_001171505.1 | 9 | 875 | |||
NM_001178036.1 | NP_001171507.1 | 10 | 1352 | |||
NM_001178037.1 | NP_001171508.1 | 9 | 918 | |||
NM_001321905.1 | NP_001308834.1 | 10 | 1378 | |||
NM_001321906.1 | NP_001308835.1 | 10 | 994 | |||
NM_001321907.1 | NP_001308836.1 | 9 | 1263 | |||
NM_001321908.1 | NP_001308837.1 | 9 | 1237 | |||
NM_001321909.1 | NP_001308838.1 | 8 | 852 | |||
NM_001321910.1 | NP_001308839.1 | 10 | 1321 | |||
NM_001321911.1 | NP_001308840.1 | 9 | 1320 | |||
NM_001321912.1 | NP_001308841.1 | 10 | 1352 | |||
NM_001321913.1 | NP_001308842.1 | 8 | 1148 | |||
NM_001321914.1 | NP_001308843.1 | 9 | 873 | |||
NM_020868.4 | NP_065919.2 | 9 | 1263 | |||
XM_017004565.1 | XP_016860054.1 | 10 | 1422 | |||
XM_017004566.1 | XP_016860055.1 | 8 | 867 | |||
GenBank mRNA | AB040925.2 | - | 9 | 830 | ||
AK289967.1 | - | 9 | 962 | |||
AK296490.1 | - | 9 | 950 | |||
AK315147.1 | - | 9 | 1067 | |||
AY172661.3 | - | 9 | 842 | |||
AY387785.1 | - | 9 | 864 | |||
BC030832.1 | - | 9 | 1299 | 4811771 | ||
DQ857321.1 | - | 9 | 830 | |||
DQ857322.1 | - | 9 | 854 | |||
DQ857323.1 | - | 9 | 821 |