Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004360.3 | NP_001004360.2 | 5 | 514 | ||
NM_001178034.1 | NP_001171505.1 | 5 | 441 | |||
NM_001178036.1 | NP_001171507.1 | 6 | 918 | |||
NM_001178037.1 | NP_001171508.1 | 5 | 484 | |||
NM_001321905.1 | NP_001308834.1 | 6 | 944 | |||
NM_001321906.1 | NP_001308835.1 | 6 | 560 | |||
NM_001321907.1 | NP_001308836.1 | 5 | 829 | |||
NM_001321908.1 | NP_001308837.1 | 5 | 803 | |||
NM_001321909.1 | NP_001308838.1 | 4 | 418 | |||
NM_001321910.1 | NP_001308839.1 | 6 | 887 | |||
NM_001321911.1 | NP_001308840.1 | 5 | 886 | |||
NM_001321912.1 | NP_001308841.1 | 6 | 918 | |||
NM_001321913.1 | NP_001308842.1 | 4 | 714 | |||
NM_001321914.1 | NP_001308843.1 | 5 | 444 | |||
NM_020868.4 | NP_065919.2 | 5 | 829 | |||
XM_017004565.1 | XP_016860054.1 | 6 | 988 | |||
XM_017004566.1 | XP_016860055.1 | 4 | 433 | |||
GenBank mRNA | AB040925.2 | - | 5 | 396 | ||
AK055256.1 | - | 7 | 1002 | |||
AK289967.1 | - | 5 | 528 | |||
AK296490.1 | - | 5 | 516 | |||
AK315147.1 | - | 5 | 633 | |||
AY172661.3 | - | 5 | 408 | |||
AY387785.1 | - | 5 | 430 | |||
BC030832.1 | - | 5 | 865 | 4811771 | ||
BI597037.1 | - | 5 | 471 | |||
DQ857321.1 | - | 5 | 396 | |||
DQ857322.1 | - | 5 | 420 | |||
DQ857323.1 | - | 5 | 387 |