Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021801.4 | NP_068573.2 | 6 | 917 | ||
XM_011520219.2 | XP_011518521.1 | 7 | 1097 | |||
GenBank mRNA | AF230354.1 | - | 6 | 901 | ||
AF248646.1 | - | 6 | 904 | |||
AF291664.1 | - | 6 | 914 | |||
AJ251531.1 | - | 6 | 911 |