Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282447.2 | NP_001269376.1 | 6 | 1182 | ||
NM_025220.4 | NP_079496.1 | 6 | 1182 | |||
NM_153202.3 | NP_694882.1 | 6 | 1182 | |||
XM_005260843.1 | XP_005260900.1 | 6 | 639 | |||
XM_006723639.1 | XP_006723702.1 | 6 | 639 | |||
XM_006723640.1 | XP_006723703.1 | 6 | 639 | |||
XM_006723644.2 | XP_006723707.1 | 6 | 639 | |||
XM_011529366.1 | XP_011527668.1 | 6 | 538 | |||
XM_011529367.1 | XP_011527669.1 | 6 | 499 | |||
XM_011529368.1 | XP_011527670.1 | 6 | 639 | |||
XM_011529370.1 | XP_011527672.1 | 6 | 639 | |||
XM_011529371.1 | XP_011527673.1 | 6 | 639 | |||
XM_011529373.1 | XP_011527675.1 | 6 | 536 | |||
XM_017028080.1 | XP_016883569.1 | 6 | 639 | |||
XM_017028081.1 | XP_016883570.1 | 6 | 560 | |||
XM_017028082.1 | XP_016883571.1 | 6 | 639 | |||
XM_017028083.1 | XP_016883572.1 | 6 | 639 | |||
GenBank mRNA | AB055891.1 | - | 6 | 583 | ||
AF466287.1 | - | 6 | 501 | |||
AK123015.1 | - | 3 | 662 | |||
AK126487.1 | - | 4 | 1438 | |||
AK126498.1 | - | 4 | 344 | |||
AK226148.1 | - | 5 | 886 | |||
AK300429.1 | - | 6 | 550 | |||
AK304037.1 | - | 6 | 536 | |||
AY358314.1 | - | 6 | 609 | |||
BC125112.1 | - | 4 | 799 | |||
BC125113.1 | - | 6 | 543 | 40118831 | ||
DB225158.1 | - | 6 | 560 |