Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278919.1 | NP_001265848.1 | 13 | 2679 | ||
NM_001278920.1 | NP_001265849.1 | 13 | 2546 | |||
NM_030779.3 | NP_110406.1 | 14 | 2787 | |||
NM_173092.2 | NP_775115.1 | 14 | 2520 | |||
XM_011525310.2 | XP_011523612.1 | 14 | 2520 | |||
XM_017025175.1 | XP_016880664.1 | 14 | 2787 | |||
XM_017025176.1 | XP_016880665.1 | 14 | 2754 | |||
XM_017025177.1 | XP_016880666.1 | 13 | 2679 | |||
XM_017025178.1 | XP_016880667.1 | 13 | 2646 | |||
XM_017025179.1 | XP_016880668.1 | 15 | 2628 | |||
XM_017025180.1 | XP_016880669.1 | 14 | 2623 | |||
GenBank mRNA | AF311913.1 | - | 14 | 2718 | ||
AK090969.1 | - | 7 | 1017 | |||
AK091877.1 | - | 14 | 2520 | |||
AK296498.1 | - | 13 | 2652 | |||
AK298363.1 | - | 13 | 2521 |