Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244606.1 | NP_001231535.1 | 4 | 353 | ||
NM_032549.3 | NP_115938.1 | 3 | 667 | |||
XM_005250630.3 | XP_005250687.1 | 5 | 435 | |||
XM_011516605.2 | XP_011514907.1 | 4 | 367 | |||
XM_011516608.2 | XP_011514910.1 | 3 | 629 | |||
XM_011516609.2 | XP_011514911.1 | 3 | 629 | |||
XM_011516611.2 | XP_011514913.1 | 3 | 629 | |||
XM_011516613.2 | XP_011514915.1 | 3 | 629 | |||
XM_017012699.1 | XP_016868188.1 | 3 | 629 | |||
XM_017012700.1 | XP_016868189.1 | 4 | 367 | |||
XM_017012701.1 | XP_016868190.1 | 4 | 318 | |||
XM_017012702.1 | XP_016868191.1 | 5 | 435 | |||
XM_017012703.1 | XP_016868192.1 | 3 | 629 | |||
XM_017012704.1 | XP_016868193.1 | 3 | 629 | |||
XM_017012705.1 | XP_016868194.1 | 4 | 405 | |||
GenBank mRNA | AF359563.1 | - | 3 | 667 | ||
AK311968.1 | - | 2 | 318 | |||
BC008497.1 | - | 4 | 318 | 4279910 |