Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NALCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052867.2 | NP_443099.1 | 3 | 445 | ||
XM_011521067.2 | XP_011519369.1 | 3 | 385 | |||
XM_011521068.2 | XP_011519370.1 | 3 | 368 | |||
XM_011521069.2 | XP_011519371.1 | 3 | 385 | |||
XM_017020534.1 | XP_016876023.1 | 3 | 416 | |||
XM_017020535.1 | XP_016876024.1 | 3 | 358 | |||
GenBank mRNA | AK094669.1 | - | 3 | 416 | ||
AY141972.1 | - | 3 | 339 | |||
BC028390.1 | - | 3 | 352 | |||
BC064343.1 | - | 2 | 284 |