Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PKD1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076780.1 | NP_001070248.1 | 5 | 857 | ||
NM_052892.3 | NP_443124.3 | 5 | 857 | |||
NR_126532.1 | 5 | 857 | ||||
GenBank mRNA | AY164483.1 | - | 5 | 857 |