Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039350.2 | NP_001034439.1 | 5, 6 | 1028 | ||
NM_001936.4 | NP_001927.3 | 5, 6 | 730 | |||
NM_130797.3 | NP_570629.2 | 5, 6 | 1036 | |||
XM_017011808.1 | XP_016867297.1 | 6, 7 | 982 | |||
XM_017011809.1 | XP_016867298.1 | 6, 7 | 745 | |||
XM_017011810.1 | XP_016867299.1 | 6, 7 | 848 | |||
XM_017011811.1 | XP_016867300.1 | 6, 7 | 772 | |||
GenBank mRNA | BC150304.1 | - | 5, 6 | 1027 | 8860401 | |
BX647966.1 | - | 5, 6 | 793 | |||
M96859.1 | - | 5, 6 | 777 | |||
M96860.1 | - | 5, 6 | 624 |