Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025096.1 | NP_001020267.1 | 7 | 1120 | ||
NM_001025097.1 | NP_001020268.1 | 7 | 1006 | |||
NM_001308188.1 | NP_001295117.1 | 8 | 1190 | |||
NM_144662.2 | NP_653263.2 | 7 | 1138 | |||
XM_011525818.1 | XP_011524120.1 | 8 | 1073 | |||
GenBank mRNA | BC021713.1 | - | 8 | 1130 | 4826048 | |
BC025389.1 | - | 7 | 1116 | |||
BC025393.1 | - | 7 | 994 | |||
BC028371.1 | - | 7 | 994 | 4828792 | ||
BC028686.1 | - | 7 | 994 | 4828931 | ||
BC042820.1 | - | 7 | 1120 | 5268996 | ||
BC047355.1 | - | 7 | 1112 |