Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144691.4 | NP_653292.2 | 17, 18 | 2195 | ||
XM_017026355.1 | XP_016881844.1 | 15, 16 | 2207 | |||
XM_017026357.1 | XP_016881846.1 | 16, 17 | 2135 | |||
XM_017026359.1 | XP_016881848.1 | 16 | 2366 | |||
XM_017026364.1 | XP_016881853.1 | 15, 16 | 1843 | |||
GenBank mRNA | AK125713.1 | - | 6 | 1606 |