Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258205.1 | NP_001245134.1 | 11 | 1779 | ||
NM_145068.3 | NP_659505.1 | 11 | 1779 | |||
GenBank mRNA | AF514998.1 | - | 11 | 1521 | ||
AJ487035.2 | - | 11 | 1779 | |||
AK127726.1 | - | 5 | 731 | |||
AK304768.1 | - | 3 | 446 | |||
AY118267.1 | - | 11 | 1589 | |||
AY118268.1 | - | 11 | 1589 | |||
BC104866.1 | - | 11 | 1551 | 8143869 | ||
BC104868.1 | - | 11 | 1551 | 8143871 | ||
BC143294.1 | - | 12 | 1720 | 9051797 | ||
BC143299.1 | - | 12 | 1720 | 9051803 | ||
BC143330.1 | - | 11 | 1551 | 9051835 | ||
BC143333.1 | - | 12 | 1640 | |||
BC143334.1 | - | 11 | 1633 | |||
BX537539.1 | - | 5 | 778 | |||
EU295940.1 | - | 10 | 1457 |