Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002405.3 | NP_002396.2 | 1, 2 | 513 | ||
GenBank mRNA | AK292123.1 | - | 1, 2 | 454 | ||
BC094814.1 | - | 1, 2 | 474 | 5930073 | ||
CR456518.1 | - | 1, 2 | 306 | |||
DA815501.1 | - | 1, 2 | 513 |