Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA2D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172364.4 | NP_758952.4 | 4 | 695 | ||
XM_011521041.2 | XP_011519343.1 | 4 | 4597 | |||
GenBank mRNA | AF516695.1 | - | 4 | 827 | ||
BC150186.1 | - | 4 | 628 | 40148321 | ||
BX537436.1 | - | 4 | 658 | |||
BX537437.1 | - | 4 | 696 |