Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U18810.1 | - | 1730 |
Gene Symbol: MTND2P28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026901.1 | - | 1968 |