Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002426.5 | NP_002417.2 | 2 | 434 | ||
GenBank mRNA | AK313959.1 | - | 2 | 383 | ||
BC112301.1 | - | 2 | 434 | 8327769 | ||
BC143773.1 | - | 2 | 434 | 9052287 | ||
BE440047.1 | - | 2 | 384 | |||
L23808.1 | - | 2 | 349 |