Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000744.6 | NP_000735.1 | 3 | 493 | ||
NM_001256573.1 | NP_001243502.1 | 3 | 406 | |||
NR_046317.1 | 3 | 493 | ||||
XM_011528524.1 | XP_011526826.1 | 1 | 37 | |||
XM_017027624.1 | XP_016883113.1 | 4 | 525 | |||
XM_017027625.1 | XP_016883114.1 | 4 | 525 | |||
GenBank mRNA | AK314907.1 | - | 3 | 290 | ||
BC096290.1 | - | 3 | 401 | 40007296 | ||
BC096291.1 | - | 3 | 209 | 40007297 | ||
BC096292.1 | - | 3 | 314 | 40007298 | ||
BC096293.3 | - | 3 | 401 | 40007299 | ||
L35901.1 | - | 3 | 440 | |||
U62433.1 | - | 3 | 493 | |||
X87629.1 | - | 2 | 187 | |||
X89743.1 | - | 1 | 257 | |||
Y08421.1 | - | 3 | 262 |