Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002448.3 | NP_002439.2 | 2 | 1136 | ||
GenBank mRNA | AK290636.1 | - | 2 | 1137 | ||
BC021285.2 | - | 2 | 1103 | 5018103 | ||
BC067353.1 | - | 2 | 896 | 4893288 | ||
M97676.1 | - | 2 | 1123 |