Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136153.1 | NP_001129625.1 | 2 | 130 | ||
NM_004381.4 | NP_004372.3 | 2 | 139 | |||
GenBank mRNA | BC008394.1 | - | 2 | 121 | ||
BC077075.1 | - | 2 | 112 | 5766821 | ||
BT007042.1 | - | 2 | 106 | |||
U31903.1 | - | 2 | 143 | |||
X98054.1 | - | 2 | 139 |
Gene Symbol: TNXB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U52696.1 | - | 2 | 110 |