Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004417.3 | NP_004408.1 | 4 | 1702 | ||
GenBank mRNA | AK298047.1 | - | 5 | 1636 | ||
AK300487.1 | - | 5 | 1616 | |||
X68277.1 | - | 4 | 1687 |