Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004853.2 | NP_004844.1 | 4 | 401 | ||
GenBank mRNA | AF036715.1 | - | 4 | 251 | ||
AF062077.1 | - | 4 | 401 | |||
AF115323.1 | - | 4 | 401 | |||
BC009713.1 | - | 4 | 274 | 3899766 | ||
BT007319.1 | - | 4 | 251 |