Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293228.1 | NP_001280157.1 | 3 | 2657 | ||
NM_001293231.1 | NP_001280160.1 | 2 | 1440 | |||
NM_001293233.1 | NP_001280162.1 | 3 | 2347 | |||
NM_005378.5 | NP_005369.2 | 3 | 2347 | |||
XM_017004168.1 | XP_016859657.1 | 2 | 2178 | |||
GenBank mRNA | AK298225.1 | - | 4 | 1640 | ||
BC002712.2 | - | 2 | 2065 | 3627330 |