Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284209.1 | NP_001271138.1 | 5 | 479 | ||
NM_001284210.1 | NP_001271139.1 | 5 | 458 | |||
NM_001284211.1 | NP_001271140.1 | 5 | 458 | |||
NM_005466.3 | NP_005457.2 | 5 | 458 | |||
GenBank mRNA | AF074723.1 | - | 5 | 429 | ||
AF155104.1 | - | 4 | 380 | |||
AK222587.1 | - | 5 | 434 | |||
AK223346.1 | - | 5 | 423 | |||
AK290647.1 | - | 5 | 423 | |||
AK300417.1 | - | 4 | 730 | |||
AK300460.1 | - | 5 | 444 | |||
AK304361.1 | - | 5 | 423 | |||
BC004106.2 | - | 5 | 432 | 3936675 | ||
BM152825.1 | - | 5 | 448 | |||
BM802752.1 | - | 410 | ||||
BT006831.1 | - | 5 | 405 | |||
CR541697.1 | - | 5 | 405 | |||
U78082.1 | - | 5 | 455 |