Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144878.1 | NP_001138350.1 | 9 | 958 | ||
NM_001144879.1 | NP_001138351.1 | 7 | 648 | |||
NM_005536.3 | NP_005527.1 | 8 | 757 | |||
GenBank mRNA | AF042729.2 | - | 8 | 728 | ||
AK297078.1 | - | 7 | 595 | |||
AK300750.1 | - | 9 | 887 | |||
AK312823.1 | - | 8 | 703 | |||
BC008381.1 | - | 8 | 723 | 3682305 | ||
BC009565.1 | - | 8 | 727 | 3682657 | ||
X66922.1 | - | 7 | 666 |