Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005567.3 | NP_005558.1 | 6 | 973 | ||
GenBank mRNA | AK055977.1 | - | 5 | 636 | ||
AK057776.1 | - | 5 | 604 | |||
AK293183.1 | - | 5 | 647 | |||
AK295442.1 | - | 5 | 401 | |||
AK301040.1 | - | 5 | 881 | |||
AK301444.1 | - | 5 | 621 | |||
AK307665.1 | - | 6 | 894 | |||
BC002403.2 | - | 6 | 906 | 2822371 | ||
BC002998.1 | - | 6 | 906 | 2822371 | ||
BC015761.1 | - | 6 | 938 | 4858122 | ||
KU178164.1 | - | 5 | 794 | |||
KU178165.1 | - | 4 | 541 | |||
L13210.1 | - | 6 | 973 | |||
X79089.1 | - | 6 | 925 |