Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304990.1 | NP_001291919.1 | 3 | 515 | ||
NM_005840.2 | NP_005831.1 | 2 | 659 | |||
GenBank mRNA | AK125483.1 | - | 2 | 584 | ||
AK289543.1 | - | 2 | 652 | |||
BC069609.1 | - | 1 | 221 | 7262284 | ||
BC074972.2 | - | 1 | 197 | 30915293 | ||
BC074973.2 | - | 1 | 197 | 30915511 | ||
DA102958.1 | - | 3 | 515 |