Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEUROG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006161.2 | NP_006152.2 | 1 | 1084 | ||
GenBank mRNA | AB593107.1 | - | 1 | 1096 | ||
BC008687.1 | - | 1 | 1084 | 3872172 | ||
BC028226.1 | - | 1 | 1084 | 5241507 |