Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF022654.1 | - | 4 | 1500 | ||
AJ002367.1 | - | 5 | 1753 | |||
AJ002368.1 | - | 4 | 1346 | |||
BC008829.2 | - | 6 | 1937 | 4122010 |