Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197237.1 | NP_001184166.1 | 8 | 2700 | ||
NM_001197239.1 | NP_001184168.1 | 6 | 2048 | |||
NM_001197240.1 | NP_001184169.1 | 7 | 2549 | |||
NM_006995.4 | NP_008926.2 | 8 | 2678 | |||
NM_181531.2 | NP_853509.1 | 7 | 2330 | |||
XM_005248797.3 | XP_005248854.1 | 8 | 2695 | |||
XM_005248798.4 | XP_005248855.1 | 7 | 2655 | |||
XM_006714953.3 | XP_006715016.1 | 8 | 2947 | |||
XM_006714955.3 | XP_006715018.1 | 8 | 2924 | |||
XM_011514228.2 | XP_011512530.1 | 8 | 3000 | |||
XM_011514229.2 | XP_011512531.1 | 8 | 2509 | |||
XM_011514231.2 | XP_011512533.1 | 6 | 2373 | |||
XM_017010166.1 | XP_016865655.1 | 8 | 2698 | |||
XM_017010167.1 | XP_016865656.1 | 7 | 2600 | |||
XM_017010168.1 | XP_016865657.1 | 8 | 3180 | |||
XM_017010169.1 | XP_016865658.1 | 8 | 2898 | |||
GenBank mRNA | BC013258.2 | - | 6 | 2250 | ||
BC014021.2 | - | 8 | 2676 | 4124138 | ||
U90550.1 | - | 8 | 2653 |