Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012333.2 | NP_001012333.1 | 5 | 599 | ||
NM_001012334.2 | NP_001012334.1 | 5 | 645 | |||
NM_001270550.1 | NP_001257479.1 | 5 | 824 | |||
NM_001270551.1 | NP_001257480.1 | 4 | 770 | |||
NM_001270552.1 | NP_001257481.1 | 4 | 360 | |||
NM_002391.4 | NP_002382.1 | 5 | 528 | |||
NR_073039.1 | 4 | 451 | ||||
XM_011520116.2 | XP_011518418.1 | 5 | 811 | |||
XM_017017764.1 | XP_016873253.1 | 5 | 667 | |||
GenBank mRNA | AK130889.1 | - | 5 | 645 | ||
AK308210.1 | - | 4 | 1416 | |||
AK309963.1 | - | 4 | 1544 | |||
BC011704.2 | - | 5 | 459 | 3609004 | ||
BF684205.1 | - | 4 | 389 | |||
BX346163.2 | - | 4 | 728 | |||
CN266354.1 | - | 4 | 413 | |||
CN266369.1 | - | 5 | 520 | |||
DQ323888.1 | - | 4 | 346 | |||
DQ323889.1 | - | 4 | 269 | |||
DQ323890.1 | - | 5 | 271 | |||
M69148.1 | - | 4 | 423 | |||
X55110.1 | - | 5 | 448 |