Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBPJL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281448.1 | NP_001268377.1 | 9 | 1015 | ||
NM_001281449.1 | NP_001268378.1 | 9 | 1015 | |||
NM_014276.3 | NP_055091.2 | 9 | 1015 | |||
XM_011528521.2 | XP_011526823.1 | 9 | 1022 | |||
XM_011528522.2 | XP_011526824.1 | 9 | 1022 | |||
XM_011528523.2 | XP_011526825.1 | 9 | 1022 | |||
GenBank mRNA | AB024964.1 | - | 9 | 935 | ||
AB026048.1 | - | 9 | 935 | |||
AB027710.1 | - | 9 | 935 |