Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168335.1 | NP_001161807.1 | 2, 3 | 375 | ||
NM_002396.4 | NP_002387.1 | 2, 3 | 375 | |||
GenBank mRNA | AK313391.1 | - | 2, 3 | 271 | ||
AU119085.1 | - | 2, 3 | 372 | |||
BC000147.1 | - | 2, 3 | 317 | 2900677 | ||
BI465083.1 | - | 2, 3 | 374 | |||
BI559761.1 | - | 2, 3 | 341 | |||
DA644731.1 | - | 2, 3 | 375 | |||
M55905.1 | - | 2, 3 | 205 |