Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007271.1 | NP_001007272.1 | 8 | 1621 | ||
NM_001007272.1 | NP_001007273.1 | 4 | 896 | |||
NM_001007273.1 | NP_001007274.1 | 6 | 1195 | |||
NM_001320842.1 | NP_001307771.1 | 7 | 1413 | |||
NM_001320843.1 | NP_001307772.1 | 5 | 991 | |||
NM_016364.3 | NP_057448.3 | 4 | 754 | |||
XM_005269883.3 | XP_005269940.1 | 7 | 1391 | |||
XM_005269884.4 | XP_005269941.3 | 4 | 1183 | |||
XM_005269887.1 | XP_005269944.1 | 7 | 1493 | |||
XM_005269890.1 | XP_005269947.1 | 5 | 1093 | |||
XM_011539853.1 | XP_011538155.1 | 8 | 1630 | |||
XM_011539854.2 | XP_011538156.1 | 6 | 1169 | |||
XM_011539855.1 | XP_011538157.1 | 4 | 872 | |||
XM_011539856.2 | XP_011538158.1 | 6 | 1173 | |||
XM_017016313.1 | XP_016871802.1 | 4 | 846 | |||
GenBank mRNA | AB027004.1 | - | 4 | 768 | ||
AB103375.1 | - | 4 | 768 | |||
AK057012.1 | - | 5 | 964 | |||
AK127886.1 | - | 7 | 1413 | |||
AY040091.1 | - | 8 | 1585 | |||
AY674051.1 | - | 8 | 1594 | |||
BC009778.1 | - | 5 | 880 | 4047963 |