Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161330.1 | NP_001154802.1 | 2 | 178 | ||
NM_018984.3 | NP_061857.3 | 2 | 178 | |||
XM_005268984.4 | XP_005269041.1 | 2 | 889 | |||
XM_005268985.2 | XP_005269042.1 | 2 | 108 | |||
XM_017019493.1 | XP_016874982.1 | 1 | 22 | |||
GenBank mRNA | AB072355.1 | - | 2 | 91 | ||
AB072356.1 | - | 2 | 91 | |||
BC062341.1 | - | 2 | 170 | 6376684 | ||
BP363810.1 | - | 2 | 178 |