Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199746.1 | NP_001186675.1 | 1 | 1070 | ||
NM_001199747.1 | NP_001186676.1 | 2 | 95 | |||
NM_019558.3 | NP_062458.1 | 1 | 1070 | |||
GenBank mRNA | BC038709.1 | - | 1 | 920 | 4614350 | |
BC090853.1 | - | 2 | 769 | 6180525 | ||
BQ429720.1 | - | 2 | 45 |