Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303034.1 | NP_001289963.1 | 2 | 188 | ||
NM_020402.3 | NP_065135.2 | 2 | 188 | |||
XM_011520234.1 | XP_011518536.1 | 1 | 3322 | |||
GenBank mRNA | AF199235.2 | - | 2 | 188 | ||
AJ278118.1 | - | 2 | 158 | |||
AJ295237.1 | - | 2 | 158 | |||
BC143303.1 | - | 2 | 171 |