Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127357.1 | NP_001120829.1 | 4 | 276 | ||
NM_001127358.1 | NP_001120830.1 | 4 | 353 | |||
NM_001127359.1 | NP_001120831.1 | 4 | 276 | |||
NM_001127360.1 | NP_001120832.1 | 4 | 353 | |||
NM_020432.4 | NP_065165.3 | 3 | 264 | |||
XM_005250508.4 | XP_005250565.1 | 5 | 328 | |||
XM_005250509.4 | XP_005250566.1 | 5 | 253 | |||
XM_006716063.3 | XP_006716126.1 | 5 | 328 | |||
XM_011516422.2 | XP_011514724.1 | 5 | 328 | |||
XM_011516423.2 | XP_011514725.1 | 5 | 328 | |||
XM_017012458.1 | XP_016867947.1 | 4 | 353 | |||
XM_017012459.1 | XP_016867948.1 | 4 | 353 | |||
GenBank mRNA | AK090643.1 | - | 4 | 332 | ||
AK225854.1 | - | 4 | 340 | |||
AK308289.1 | - | 4 | 305 | |||
AL136883.1 | - | 4 | 244 | |||
AL831969.2 | - | 5 | 328 | |||
BC022419.1 | - | 4 | 276 | |||
BC067742.1 | - | 4 | 257 | |||
BC127093.1 | - | 4 | 192 | 40123852 | ||
BC127094.1 | - | 3 | 180 | 40123853 | ||
BP321440.1 | - | 4 | 373 |