Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002423.4 | NP_002414.1 | 1, 2 | 169 | ||
GenBank mRNA | AK222980.1 | - | 1, 2 | 169 | ||
AK293359.1 | - | 1, 2 | 163 | |||
AK309687.1 | - | 1, 2 | 166 | |||
BC003635.2 | - | 1, 2 | 147 | 3545760 | ||
BP332706.1 | - | 1, 2 | 163 | |||
X07819.1 | - | 1, 2 | 143 | |||
Z11887.1 | - | 1, 2 | 143 |