Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056567.1 | - | 4 | 2400 | ||
AK094200.1 | - | 3 | 1772 | |||
BC015180.1 | - | 3 | 2254 | 3905733 |